FHT 6020 Area Monitor Display and Alarms
Actual product may vary
Thermo Scientific™

FHT 6020 Area Monitor Display and Alarms

Increase nuclear power plant safety and emergency preparedness with an environmental monitoring solution to identify and assess the level of gamma and or neutron radiation.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDescription
HSD32423Radiation Detection Alarm, Vehicle UnitRS485 PC port, No ports for FH40G probes, 2/2 analog IN/OUT, Open cable
4255909Radiation Detection Alarm, US Alarm Unit SetUS Alarm Unit Set (Comprising FHT 6025N)
HSD-3-2-4-0-0Radiation Detection Alarm, Network UnitRS232 PC Port, Powered by FHT 6025N alarm unit, 2 ports for FH40G probes, No analog IN/OUT, No connection cable
HSA34423USRadiation Detection Alarm, Standalone UnitRS485 PC port, 2 ports for FH40G, 2/2 analog IN/OUT, Integrated AC Power Adapter
HSA04400USRadiation Detection Alarm, Network UnitRS485 PC port, AC adapter, No ports for FH40G probes, No analog IN/OUT, Mains cable
HSA32423USRadiation Detection Alarm, Standalone UnitRS232 PC Port, 2 ports for FH40G, 2/2 analog IN/OUT, Integrated AC Power Adapter
HSF32400Radiation Detection Alarm, Vehicle UnitRS485 PC port, 2 ports for FH40G probes, No analog IN/OUT, Open cable
HSA32400USRadiation Detection Alarm, Standalone UnitRS232 PC Port, 2 ports for FH40G, No analog IN/OUT
HSA34400USRadiation Detection Alarm, Network UnitRS485 PC port, AC adapter, 2 ports for FH40G probes, No analog IN/OUT, Mains cable
HSF32423Radiation Detection Alarm, Standalone UnitRS232 PC Port, 2 ports for FH40G, 2/2 analog IN/OUT, connection for external DC incl. DC filter
HSD02400Radiation Detection Alarm, Network UnitRS232 PC Port, Powered by FHT 6025N alarm unit, No ports for FH40G probes, No analog IN/OUT, No connection cable
HSD34400Radiation Detection Alarm, Network UnitRS485 PC port, Powered by FHT 6025N alarm unit, 2 ports for FH40G probes, No analog IN/OUT, No connection cable
HSD34423Radiation Detection Alarm, Network UnitRS485 PC port, Powered by FHT 6025N alarm unit, 2 ports for FH40G probes, 2/2 analog IN/OUT, No connection cable
425590601Connection Cable for Radiation Detection AlarmConnection Cable (2 m)
HSA02400USRadiation Detection Alarm, Standalone UnitRS232 PC Port, No analog IN/OUT
HSD04400Radiation Detection Alarm, Network UnitRS485 PC port, Powered by FHT 6025N alarm unit, No ports for FH40G probes, No analog IN/OUT, No connection cable
HSF02400Radiation Detection Alarm, Vehicle UnitRS485 PC port, No ports for FH40G probes, No analog IN/OUT, Open cable
Catalog number HSD32423
Price (USD)
-
Product Type:
Radiation Detection Alarm, Vehicle Unit
Description:
RS485 PC port, No ports for FH40G probes, 2/2 analog IN/OUT, Open cable

Nuclear facilities need accurate, reliable, sustained early warning of radiation leaks. The Thermo Scientific™ FHT 6020 Area Monitor Display and Alarms offers maximum reliability for permanent operation within nuclear facilities or environmental networks with up to 16 measurement channels that can process and display data from Thermo Scientific intelligent dose rate probes, FH 40 G series probes as well as analog or digital input. All channels can be operated simultaneously, along with audible and visible alarms, to measure and assess the level of gamma and neutron radiation.

The FHT 6020 Area Monitor Display is an industry-proven configurable area monitor capable of stand-alone or network operation.

Measurement Channels

  • Total number of measurement channels: 16
  • Maximum number of FH40G probe channels: 2
  • Maximum number of analog input channels: 2
  • Maximum number of digital input channels: 4

Measurement History

  • Ring buffer storage for > 1000 data entries
  • Measurement memory for probes of the FH40G range

PC/Network Interface (selectable at order)

  • Connection to a local PC or communication adapters (e.a serial to TCP/IP) via RS-232 interface
  • Incorporation in measuring networks via RS 485 interface

Power Supply Options (selectable at order)

  • External DC supply (typ. 24V)
  • International wide range AC
  • Car supply via special DC filter (9V)

Output Channels

  • Maximum number of analog output channels: 2
  • Maximum number of digital output channels: 5

Firmware

  • Equipped with a flash EEPROM storing the firmware, allowing updates
  • Customer-specific programs possible on request

Software

  • Easily and conveniently configured with a PC via serial interface, and through the configuration program FHT 6020.exe.
  • Networking of multiple units and further measurement equipment via Thermo Scientific™ NETVIEW or Thermo Scientific ™VIEWPOINT Software Packages
Specifications
DescriptionRS485 PC port, No ports for FH40G probes, 2/2 analog IN/OUT, Open cable
Product TypeRadiation Detection Alarm, Vehicle Unit
Unit SizeEach