Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™

Have Questions?
Catalog NumberQuantityPackaging
J-7620-17
also known as J/7620/17
2.5 LHDPE plastic bottle
Catalog number J-7620-17
also known as J/7620/17
Price (USD)
-
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point100°C
CAS1310-73-2
ChEBICHEBI:32145
ColorColorless
IUPAC Namesodium;hydroxide
View more
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp