You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
Shop All Products
Chemicals
Salts and Inorganics
Inorganic Salts
Alkali Metal Salts
EDTA Tetrasodium Salt Dihydrate, 99-101%, Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
EDTA Tetrasodium Salt Dihydrate, 99-101%, Ultrapure, Thermo Scientific Chemicals
Catalog number:
J15700.C4
25 kg, Plastic bottle, Each
Certificates
Thermo Scientific Chemicals
EDTA Tetrasodium Salt Dihydrate, 99-101%, Ultrapure, Thermo Scientific Chemicals
Catalog number:
J15700.C4
25 kg, Plastic bottle, Each
Quantity
1 kg
25 kg
Packaging
Plastic bottle
Certificates
Catalog number:
J15700.C4
Price (USD)
Contact Us ›
Request bulk or custom quote
Save to list
Technical Support
Customer Service
Chemical Identifiers
CAS
10378-23-1
IUPAC Name
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
Molecular Formula
C10H20N2Na4O10
InChI Key
DGWOWDJDIBPYOR-UHFFFAOYSA-N
SMILES
O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View More
Specifications
Specification Sheet
Form
White crystalline powder
pH
(1%, H₂O): 10.7 to 11.7
Assay (unspecified)
99 to 101% (as is)
Identification (FTIR)
Conforms
pH
(5%, H₂O): 10.7 to 12.0
Description
RUO – Research Use Only
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search