Thermo Scientific Chemicals

myo-Inositol, 99%, Thermo Scientific Chemicals

Catalog number: J60828.36
500 g, Each
Thermo Scientific Chemicals

myo-Inositol, 99%, Thermo Scientific Chemicals

Catalog number: J60828.36
500 g, Each
Quantity
Catalog number: J60828.36
also known as J60828-36
Price (USD)

Chemical Identifiers

CAS
87-89-8
IUPAC Name
cyclohexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H12O6
InChI Key
CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES
OC1C(O)C(O)C(O)C(O)C1O
Form
Powder
Comment
Non-animal origin
Appearance (Color)
White
Assay (GC)
98.5% min.
Loss on Drying
0.5% max.

Description

A lipotropic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text