Thermo Scientific Chemicals

Agarose Gel Loading Dye (6X, Alkaline)

Catalog number: J62157.AB
10 mL, Each
Thermo Scientific Chemicals

Agarose Gel Loading Dye (6X, Alkaline)

Catalog number: J62157.AB
10 mL, Each
Quantity
Catalog number: J62157.AB
also known as J62157-AB
Price (USD)

Description

Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

Solubility
Fully miscible in water.

Notes
300mM sodium hydroxide, 6mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text