Thermo Scientific Chemicals

tert-Butyl cyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: L00123.06
5 g, Each
Thermo Scientific Chemicals

tert-Butyl cyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: L00123.06
5 g, Each
Quantity
Catalog number: L00123.06
also known as L00123-06
Price (USD)

Chemical Identifiers

CAS
1116-98-9
Form
Liquid or viscous liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to white to yellow
Refractive Index
1.4180-1.4220 @ 20?C

Description

tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text