Thermo Scientific Chemicals

Vinyltriphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L00708.14
25 g, Each
Thermo Scientific Chemicals

Vinyltriphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L00708.14
25 g, Each
Quantity
Catalog number: L00708.14
also known as L00708-14
Price (USD)

Chemical Identifiers

CAS
5044-52-0
IUPAC Name
ethenyltriphenylphosphanium bromide
Molecular Formula
C20H18BrP
InChI Key
VRAYVWUMBAJVGH-UHFFFAOYSA-M
SMILES
[Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream or pale yellow
Melting Point (clear melt)
180.0-187.0?C
Form
Powder
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text