2,4-Nonanedione, 98+%
2,4-Nonanedione, 98+%
2,4-Nonanedione, 98+%
2,4-Nonanedione, 98+%
Thermo Scientific Chemicals

2,4-Nonanedione, 98+%

CAS: 6175-23-1 | C9H16O2 | 156.225 g/mol
Have Questions?
Catalog NumberQuantity
L01151.06
also known as L01151-06
5 g
Catalog number L01151.06
also known as L01151-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6175-23-1
IUPAC Namenonane-2,4-dione
Molecular FormulaC9H16O2
InChI KeyKFBXUKHERGLHLG-UHFFFAOYSA-N
SMILESCCCCCC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4585-1.4625 @ 20?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only