Thermo Scientific Chemicals

tert-Butyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L02668.06
5 g, Each
Thermo Scientific Chemicals

tert-Butyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L02668.06
5 g, Each
Quantity
Catalog number: L02668.06
also known as L02668-06
Price (USD)

Chemical Identifiers

CAS
590-42-1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
>96.0%
Refractive Index
1.4785-1.4835 @ 20?C

Description

tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text