Thermo Scientific Chemicals

2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: L05597.09
10 g, Each
Thermo Scientific Chemicals

2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: L05597.09
10 g, Each
Quantity
Catalog number: L05597.09
also known as L05597-09
Price (USD)

Chemical Identifiers

CAS
22545-15-9
IUPAC Name
2-(4-ethoxyphenyl)ethan-1-ol
Molecular Formula
C10H14O2
InChI Key
CNMVSNTVPZWQMI-UHFFFAOYSA-N
SMILES
CCOC1=CC=C(CCO)C=C1
Melting Point (clear melt)
38.5-44.5?C
Appearance (Color)
White
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or flakes

Description

2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text