Thermo Scientific Chemicals

Di-n-hexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L06326.AE
100 mL, Each
Thermo Scientific Chemicals

Di-n-hexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L06326.AE
100 mL, Each
Quantity
Catalog number: L06326.AE
also known as L06326-AE
Price (USD)

Chemical Identifiers

CAS
143-16-8
IUPAC Name
dihexylamine
Molecular Formula
C12H27N
InChI Key
PXSXRABJBXYMFT-UHFFFAOYSA-N
SMILES
CCCCCCNCCCCCC
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Refractive Index
1.4315-1.4355 @ 20?C
Form
Liquid

Description

Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text