Thermo Scientific Chemicals

Diphenyl sulfone, 99+%, Thermo Scientific Chemicals

Catalog number: L08232.30
250 g, Each
Thermo Scientific Chemicals

Diphenyl sulfone, 99+%, Thermo Scientific Chemicals

Catalog number: L08232.30
250 g, Each
Quantity
Catalog number: L08232.30
also known as L08232-30
Price (USD)

Chemical Identifiers

CAS
127-63-9
IUPAC Name
(benzenesulfonyl)benzene
Molecular Formula
C12H10O2S
InChI Key
KZTYYGOKRVBIMI-UHFFFAOYSA-N
SMILES
O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Identification (FTIR)
Conforms
Assay (GC)
≥99.0%
Form
Crystalline powder or powder
Melting Point (clear melt)
122-131?C

Description

It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text