Thermo Scientific Chemicals

Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane, Thermo Scientific Chemicals

Catalog number: L08269.AE
100 mL, Each
Thermo Scientific Chemicals

Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane, Thermo Scientific Chemicals

Catalog number: L08269.AE
100 mL, Each
Quantity
Catalog number: L08269.AE
also known as L08269-AE
Price (USD)

Chemical Identifiers

CAS
15405-86-4
Appearance (Color)
Clear to turbid colorless to yellow
Form
Liquid
Assay (Non-aqueous acid-base Titration)
0.16M ± 0.05M
Comment
May form sediment on storage

Description

Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text