3',5'-Dichloro-2'-hydroxyacetophenone, 99%
3',5'-Dichloro-2'-hydroxyacetophenone, 99%
3',5'-Dichloro-2'-hydroxyacetophenone, 99%
Thermo Scientific Chemicals

3',5'-Dichloro-2'-hydroxyacetophenone, 99%

CAS: 3321-92-4 | C8H6Cl2O2 | 205.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10006.14
also known as L10006-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3321-92-4
IUPAC Name1-(3,5-dichloro-2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H6Cl2O2
InChI KeyCJFYGRLJDKWMDI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(Cl)=CC(Cl)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Appearance (Color)White to yellow
Melting Point (clear melt)94.5-98.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only