Thermo Scientific Chemicals

L-2,4-Diaminobutyric acid dihydrochloride, 98+%, may cont. up to ca 10% monohydrochloride, Thermo Scientific Chemicals

Catalog number: L10093.03
1 g, Each
Thermo Scientific Chemicals

L-2,4-Diaminobutyric acid dihydrochloride, 98+%, may cont. up to ca 10% monohydrochloride, Thermo Scientific Chemicals

Catalog number: L10093.03
1 g, Each
Quantity
Catalog number: L10093.03
also known as L10093-03
Price (USD)

Chemical Identifiers

CAS
1883-09-6
IUPAC Name
dihydrogen 2,4-diaminobutanoic acid dichloride
Molecular Formula
C4H12Cl2N2O2
InChI Key
CKAAWCHIBBNLOJ-UHFFFAOYNA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCC(N)C(O)=O
Form
Powder
Optical Rotation
+19 ? 3? (c=1 in 5N HCl)
Assay (Titration ex Chloride)
≥98.0%
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text