Thermo Scientific Chemicals

N-(2-Hydroxyphenyl)thiourea, 97%, Thermo Scientific Chemicals

Catalog number: L10519.03
1 g, Each
Thermo Scientific Chemicals

N-(2-Hydroxyphenyl)thiourea, 97%, Thermo Scientific Chemicals

Catalog number: L10519.03
1 g, Each
Quantity
Catalog number: L10519.03
also known as L10519-03
Price (USD)

Chemical Identifiers

CAS
1520-26-9
IUPAC Name
(2-hydroxyphenyl)thiourea
Molecular Formula
C7H8N2OS
InChI Key
VYBCFZXLXJUFPM-UHFFFAOYSA-N
SMILES
NC(=S)NC1=CC=CC=C1O
Appearance (Color)
White to cream to brown
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text