Thermo Scientific Chemicals

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals

Catalog number: L18426.06
5 g, Each
Thermo Scientific Chemicals

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals

Catalog number: L18426.06
5 g, Each
Quantity
Catalog number: L18426.06
also known as L18426-06
Price (USD)

Chemical Identifiers

CAS
59084-16-1
IUPAC Name
1-acetylpiperidine-4-carbonyl chloride
Molecular Formula
C8H12ClNO2
InChI Key
OHCPVLJEAHBMEG-UHFFFAOYSA-N
SMILES
CC(=O)N1CCC(CC1)C(Cl)=O
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Appearance (Color)
White to cream to orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text