Thermo Scientific Chemicals

Quinoline-8-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L19642.06
5 g, Each
Thermo Scientific Chemicals

Quinoline-8-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L19642.06
5 g, Each
Quantity
Catalog number: L19642.06
also known as L19642-06
Price (USD)

Chemical Identifiers

CAS
38707-70-9
IUPAC Name
quinoline-8-carbaldehyde
Molecular Formula
C10H7NO
InChI Key
OVZQVGZERAFSPI-UHFFFAOYSA-N
SMILES
O=CC1=C2N=CC=CC2=CC=C1
Appearance (Color)
Pale brown to brown
Form
Powder
Assay (GC)
≥96.0%
Free acid (titration)
≤1.5%
Melting Point (clear melt)
91.5-97.5?C

Description

Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text