Histology Embedding Cassettes
LabServ™
Histology Embedding Cassettes
Histology Embedding Cassettes are disposable, made from acetal, and available in a wide range of colors for easy identification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
LBSHEC104P
also known as HEC104P
Pink4000/Cs.
LBSHEC108LG
also known as HEC108LG
Lime Green4000/Cs.
LBSHEC120PE
also known as HEC120PE
Peach4000/Cs.
LBSHEC101W
also known as HEC101W
White4000/Cs.
Catalog number LBSHEC104P
also known as HEC104P
Price (USD)
-
Color:
Pink
Quantity:
4000/Cs.
Histology Embedding Cassettes are disposable, made from acetal, and available in a wide range of colors for easy identification. The cassettes have a writing surface angle of 45° and are resistant to histology solvents.
  • Available in a wide range of colors
  • Resistant to all solvents used in histology
  • Cartons of 4000 are packaged in inner bags of 1000
  • Dimensions: 40.9 x 28.3 x 5.9 mm
  • Write surface angle of 45°
Specifications
Quantity4000/Cs.
Writing Face Angle45°
ColorPink
Unit SizeEach