Biological Shipper Packs
LabServ™

Biological Shipper Packs

For use when shipping infectious substances
Have Questions?
Catalog NumberTypeQuantity
LBSLABPAK-1
also known as LABPAK-1
Complete Shipper Kit12/Pk.
Catalog number LBSLABPAK-1
also known as LABPAK-1
Price (USD)
-
Type:
Complete Shipper Kit
Quantity:
12/Pk.
Biological Shipper Packs are complete kits for transporting biological hazards that are classified as dangerous goods. This full kit comes with outer carton, hazard labels, leak-proof inner container, screwcap lid, and an inner polystyrene block. Any additional components or accessories are sold separately, please contact customer service for any products not listed below.
  • Biological shipper complete for infectious substances
  • Meets all IATA regulatory requirements
  • Replacement components available
  • Inner Dimensions (W x H): 82 x 178 mm
  • UN Compliance: 4G Class 6.2/14. NZCAA approval number: NZ/CAA 38281-2. NZCAA approval number: NZ/CAA 38281-2
Specifications
DescriptionInfectious Substance Shipper
For Use With (Equipment)Biological Shipper Pk.
Quantity12/Pk.
Dimensions (LxWxH)82 mm x 178 mm
TypeComplete Shipper Kit
Unit SizeEach