MIDAS Gas Detector Transmitter and Sensors
MIDAS Gas Detector Transmitter and Sensors
Honeywell

MIDAS Gas Detector Transmitter and Sensors

Bringing new visibility, reliability and ease of use to gas detection in semiconductor processing and industrial manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberComponents Measured
MIDAS-E-HALChlorine, Fluorine
MIDAS-E-H2SH2S (low range)
MIDAS-E-HFXBF3, HF, NF3, WF6
MIDAS-E-LELH2, CH4, C3H6, C8H16
MIDAS-E-NH3Ammonia
MIDAS-E-SHXSiH4, Si2H6
MIDAS-E-SO2Sulfur Dioxide
MIDAS-T-004Universal transmitter only
MIDAS-E-HSHH2S (high range)
Catalog number MIDAS-E-HAL
Price (USD)
-
Components Measured:
Chlorine, Fluorine
The Midas Gas Detector uses reliable sensor technology to detect many key toxic, ambient and flammable gases in a plant. The device monitors points up to 30 m away while using patented technology to regulate flow rates and ensure error-free gas detection. The Midas Gas Detector is equipped with bright LED lights and an intuitive, interactive interface that provides instant alerts to gas readings or alarm levels. Compact in size with an easy-to-handle metal chassis, the Midas Gas Detector is simple to install - important in crowded, complex process environments where space is at a premium.
  • Choose the Midas Gas Detector to detect more than 35 gases — easily, quickly and error-free.
  • All with extended sensor calibration periods patented diagnostics and lower cost of ownership.
  • The Midas Gas Detector uses reliable sensor technology to detect many key toxic, ambient, and flammable gases in a plant.
  • The device monitors point up to 30m away while using patented technology to regulate flow rates and ensure error-free gas detection.
  • Improved visibility and ease of use.
  • The Midas Gas Detector is equipped with bright LED lights and an intuitive, interactive interface that provides instant alerts to gas readings or alarm levels.
  • The interface also includes password-protected menus for configuration, test, and calibration.
  • Easy installation. Compact in size with an easy-to-handle metal chassis, the Midas Gas Detector is simple to install. That' especially important in crowded, complex process environments where space is at a premium.
  • Numerous applications. With feature-rich capabilities that are easy to use, the Midas Gas Detector is suitable for many industry applications, including semiconductor processing, light industrial manufacturing, university laboratories, aerospace, wastewater and more.
  • Plug-and-play gas detection With flexible features and options, the Midas Gas Detector works right out of the box and configures easily to meet your needs. Each plug-and-play sensor cartridge is pre-calibrated, so it's ready when you are. That means longer sensor life, quick and easy replacement, and no operator error due to incorrect or depleted sensors. The standard power supply and communications capabilities include three onboard relays, 0-22 mA analog output and Modbus/TCP Ethernet digital outputs. Plus, the Midas Gas Detector includes the innovative Power over Ethernet (PoE) protocol — a single Ethernet connection for all power, control and communication requirements. Thanks to the Midas architecture, cost-effective integration with PLC and Fieldbus systems is greatly simplified.

Features:

  • Over 35 gases available with extended 2 year warranty
  • Smart sensor cartridge with on board 'e-calibration' certificate
  • 3 built in relays (Form C) for Alarm 1, 2 and Fault
  • Power over Ethernet (PoE) communication
  • Keypad interface and remote web browser interface
  • Optional Midas Pyrolyzer module that connects to any Midas Gas Detector
  • Password protected menu levels to preserve system integrity
  • Bright LEDs and intuitive icon driven, large multicolor backlit LCD display
  • Factory-calibrated sensors to reduce need for frequent gas testing
  • Isolated 0-22 mA analog output with fault and event reporting
  • Modbus/TCP Ethernet for easy connectivity to all control and alarm systems ports for control connectivity and service interaction
  • Robust extractive pump system (2 year lifetime) sampling up to 100 feet
  • CE marked for total immunity to Radio Frequency Interference
  • Reflex™ sensor health check for improved safety
  • TempraSureTM temperature compensation technology as appropriate
  • Event logging to review sensor history
Specifications
DescriptionMIDAS Sensor Cartridge
Measurement Range0 to 2 ppm (Cl2), 0 to 4 ppm (F2)
Warranty2-years
Components MeasuredChlorine, Fluorine
For Use With (Equipment)Midas Gas Detector
Unit SizeEach