Enteral ENFit Nutricare Feeding Tube with Guidewire and Safety Literature
GBUK Enteral Ltd
Enteral ENFit Nutricare Feeding Tube with Guidewire and Safety Literature
Enteral ENFit Nutricare Feeding Tube is a fully radiopaque, soft polyurethane tube with clear numbered cm markings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
NC10.092ISOSAF10 FR
NC08.092ISOSAF8 FR
NC12.092ISOSAF12 FR
Catalog number NC10.092ISOSAF
Price (USD)
-
Size:
10 FR
Enteral ENFit Nutricare Feeding Tube is a fully radiopaque, soft polyurethane tube with clear numbered cm markings.
  • Fitted with the ENFit connector
  • Long Term - use up to 90 days
  • Two non-opposing outlet holes
  • Atraumatic rounded tip
  • Individually wrapped
  • Latex and DEHP free
  • 5/box
  • Each pack includes the feeding tube with guidewire, fixation plaster, record of insertion sticker and safety literature
Specifications
Length (Metric)92 cm
Size10 FR
SterilitySterile
Unit SizeEach