Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™
Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™
Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™
Fisher Chemical™

Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™

CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L
Have Questions?
Catalog NumberQuantity
P-4120-53
also known as P/4120/53
500 g
Catalog number P-4120-53
also known as P/4120/53
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS584-08-7
ColorWhite
Formula Weight138.21g/mol
IUPAC Namedipotassium carbonate
InChI KeyBWHMMNNQKKPAPP-UHFFFAOYSA-L
View more
CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L