Liquid To Gas Interface, 20 Mpa (3000 Psi)
Fluke™
Liquid To Gas Interface, 20 Mpa (3000 Psi)
Fluke P5523 Liquid to Gas Media Separator. For high pressure pneumatic calibrations this unit interfaces with hydraulic deadweight testers providing a rateless liquid-to gas separation. Driven by a pneumatic pressure source (nitrogen bottle) and controlled by needle valves, high pressure gas.
Have Questions?
Catalog number P5523-20M
Price (USD)
-
Fluke P5523 Liquid to Gas Media Separator. For high pressure pneumatic calibrations this unit interfaces with hydraulic deadweight testers providing a rateless liquid-to gas separation. Driven by a pneumatic pressure source (nitrogen bottle) and controlled by needle valves, high pressure gas is balanced against the liquid in a hydraulic deadweight tester through a series of fluid traps.
Fluke P5523 Liquid to Gas Media Separator
For high pressure pneumatic calibrations this unit interfaces with hydraulic deadweight testers providing a rateless liquid-to gas separation. Driven by a pneumatic pressure source (nitrogen bottle) and controlled by needle valves, high pressure gas is balanced against the liquid in a hydraulic deadweight tester through a series of fluid traps.
Key features:
  • Pressure range 0 to 3,000 psi (210 bar)
  • For high pressure pneumatic calibrations this unit interfaces with hydraulic deadweight testers providing a rateless liquid-to gas separation. Driven by a pneumatic pressure source (nitrogen bottle) and controlled by needle valves, high pressure gas is balanced against the liquid in a hydraulic deadweight tester through a series of fluid traps.
  • * Note---since fluid mist may be transferred during operation we do not recommend this device for oxygen safe instruments using an oil operated deadweight tester.
Specifications
For Use With (Equipment)High Pressure Pneumatic Calibration
Product TypeLiquid To Gas Interface
Unit SizeEach