PathoProof™ Mastitis Major-4.2 Kits
Thermo Scientific™

PathoProof™ Mastitis Major-4.2 Kits

Identify four of the main contagious mastitis-causing microorganisms from bovine milk in just four hours using real-time PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)No. of Tests
PF0942SPathoProof Mastitis Major – 4.2 KitMust be ordered together with PF0942SB qPCR Reagents.50 Tests
PF0942LPathoProof Mastitis Major – 4.2 KitMust be ordered together with PF0942LB qPCR Reagents.384 Tests
PF0942LBqPCR reagents for use with PF0942L and PF0942LKFMust be ordered together with PF0942L or PF0942LKF.384 Tests
PF0942SBqPCR reagents for use with PF0942SMust be ordered together with PF0942S.50 Tests
Catalog number PF0942S
Price (USD)
-
Description:
PathoProof Mastitis Major – 4.2 Kit
For Use With (Equipment):
Must be ordered together with PF0942SB qPCR Reagents.
No. of Tests:
50 Tests
Accurately and rapidly identify four of the main contagious mastitis-causing organisms using Thermo Scientific™ PathoProof™ Mastitis Major-4.2 Kits. Obtain results within four hours and identify the particular pathogen causing the infection by testing either fresh or preserved milk samples. Make faster decisions on treatment and reduce the time that an infective agent has to spread in a herd.

PathoProof Mastitis Assays identify the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci. These microorganisms are responsible for 95% of all clinical and subclinical mastitis cases worldwide.
• Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample
• Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples
• All targeted species are detected simultaneously, overcoming the common problem of mixed growth results of bacterial culturing
• Validation studies indicate superior analytical specificity and sensitivity over traditional methods, even for samples that show “no growth” in culture
• Accurate detection of viable, dead and growth-inhibited microorganisms reduces 'no-growth' results

qPCR Reagents for PathoProof Mastitis Complete Assays

• Order item numbers PF1600SB and PF1600LB respectively
• Order item numbers PF0870SB and PF0870LB respectively

qPCR Reagents for PathoProof Mastitis Major Assays
• Order item numbers PF0941SB and PF0941LB respectively
• Order item numbers PF0942SB and PF0942LB respectively
• Order item numbers PF0914SB and PF0914LB respectively

PathoProof qPCR reagents are not sold separately.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Permissible use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
Specifications
DescriptionPathoProof Mastitis Major – 4.2 Kit
For Use With (Equipment)Must be ordered together with PF0942SB qPCR Reagents.
Kit TypeMajor 4.2
No. of Tests50 Tests
Product TypeReal Time PCR Reagents
For Use With (Application)Food & Beverage
GC-Rich PCR PerformanceMedium
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
All necessary reagents (except ethanol) for DNA extraction from milk. Varies by component; qPCR reagents should be stored at -20°C protected from light