PathoProof™ Complete-16 Kits for Applied Biosystems™ 7500 and 7500 Fast qPCR systems
Actual product may vary
Thermo Scientific™

PathoProof™ Complete-16 Kits for Applied Biosystems™ 7500 and 7500 Fast qPCR systems

Identify 15 mastitis-causing microorganisms from bovine milk in less than four hours, using real time qPCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)No. of Tests
PF1650LKFPathoProof Complete-16 KFFor use with the KingFisher Flex system. Must be ordered together with PF1650LB qPCR Reagents.384 Tests
PF1650SPathoProof Complete-16 kitMust be ordered together with PF1650SB qPCR Reagents50 Tests
PF1650SBqPCR reagents for use with PF1650SFor use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF1650S.50 Tests
PF1650LPathoProof Complete-16 kitMust be ordered together with PF1650LB qPCR Reagents.384 Tests
PF1650LBqPCR reagents for use with PF1650L and PF1650LKFFor use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF1650L or PF1650LKF.384 Tests
Catalog number PF1650LKF
Price (USD)
-
Description:
PathoProof Complete-16 KF
For Use With (Equipment):
For use with the KingFisher Flex system. Must be ordered together with PF1650LB qPCR Reagents.
No. of Tests:
384 Tests
Accurately and rapidly identify 15 of the main mastitis-causing microorganisms (responsible for 95% of all clinical and subclinical mastitis cases) and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci using Thermo Scientific™ PathoProof™ Complete-16 Kits for the Applied Biosystems™ 7500 and 7500 Fast qPCR systems. Obtain results within four hours and identify the particular pathogen causing the infection by testing either fresh or preserved milk samples. Make faster decisions on treatment and reduce infective agent spread in a herd.

Good-quality raw milk must meet accepted standards for chemical composition and purity as well as levels of a variety of mastitis-causing microorganisms. Identify 15 of the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci within four hours using PathoProof Complete-16 multiplex PCR. All targeted species are detected simultaneously, overcoming the common problem of mixed growth results in bacterial culturing.

  • Ensure milk and milk products are safe and suitable for their intended uses
  • Remove affected cows from the herd to ensure optimal milk quality
  • Enables faster decisions on treatment
  • Reduces time available for infective agents to spread to healthy herd mates
  • Reduces broad spectrum antibiotic treatment
  • Rapid treatment shortens inflammation time

Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample. Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples, and bulk tank milk samples.

Achieve high sensitivity, even for samples that show no growth in culture. Accurate detection of viable, dead, and growth-inhibited microorganisms reduces no-growth results.

PathoProof Complete-16 Kit identifies:

  1. Staphylococcus aureus
  2. Staphylococcus spp. (including all major coagulase-negative staphylococci)
  3. Streptococcus agalactiae
  4. Streptococcus dysgalactiae
  5. Streptococcus uberis
  6. Escherichia coli
  7. Enterococcus spp. (including E. faecalis and E. faecium)
  8. Klebsiella oxytoca and K. pneumoniae
  9. Serratia marcescens
  10. Corynebacterium bovis
  11. Arcanobacter pyogenes and Peptoniphilus (Peptostreptococcus) indolicus
  12. Staphylococcal β-lactamase gene (penicillin-resistance gene)
  13. Mycoplasma bovis
  14. Mycoplasma spp.
  15. Yeast
  16. Prototheca spp

Automated sample preparation
Significantly reduce the hands-on time needed when processing larger numbers of samples with the semi-automated Thermo Scientific™ KingFisher™ Flex instrument for magnetic bead-based DNA purification and extraction, along with PathoProof Mastitis Complete-16 KingFisher Kits and qPCR reagents.

Not all products are available for sale in all territories. Please inquire.

Permissible Use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence, or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
For testing of Food and Environmental samples only.
Specifications
DescriptionPathoProof Complete-16 KF
For Use With (Equipment)For use with the KingFisher Flex system. Must be ordered together with PF1650LB qPCR Reagents.
Kit TypeComplete-16 Kits
No. of Tests384 Tests
Product TypeqPCR Reagents for Microbial Detection
Detection MethodqPCR
For Use With (Application)Food & Beverage
GC-Rich PCR PerformanceHigh
PCR MethodMultiplex qPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
All necessary reagents (except ethanol) for DNA extraction from milk