PathoProof™ Majorû3 Kits for Applied Biosystems™ 7500 and 7500 Fast qPCR systems
Actual product may vary
Thermo Scientific™

PathoProof™ Majorû3 Kits for Applied Biosystems™ 7500 and 7500 Fast qPCR systems

Identify three of the main contagious mastitis-causing microorganisms from bovine milk in just four hours using real-time qPCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)No. of Tests
PF3050LBqPCR reagents for use with PF3050L and PF3050LKFFor use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF3050L or PF3050LKF.384 Tests
PF3050SPathoProof Major-3 kitMust be ordered together with PF3050SB qPCR Reagents.50 Tests
PF3050SBqPCR reagents for use with PF3050SFor use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF3050S.50 Tests
PF3050LPathoProof Major-3 kitMust be ordered together with PF3050LB qPCR Reagents.384 Tests
Catalog number PF3050LB
Price (USD)
-
Description:
qPCR reagents for use with PF3050L and PF3050LKF
For Use With (Equipment):
For use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF3050L or PF3050LKF.
No. of Tests:
384 Tests
Accurately and rapidly identify three of the main mastitis-causing microorganisms using Thermo Scientific™ PathoProof™ Major-3 Kits for the Applied Biosystems™ 7500 and 7500 Fast qPCR systems. Obtain results within four hours and identify the particular pathogen causing the infection by testing either fresh or preserved milk samples. Make faster decisions on treatment and reduce the time that an infective agent has to spread in a herd.

Good-quality raw milk must meet accepted standards for chemical composition and purity as well as levels of a variety of mastitis-causing microorganisms. Identify three of the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci using PathoProof Major–3 multiplex PCR. All targeted species are detected simultaneously, overcoming the common problem of mixed growth results in bacterial culturing.

  • Ensure milk and milk products are safe and suitable for their intended uses
  • Remove affected cows from the herd to ensure optimal milk quality
  • Enables faster decisions on treatment
  • Reduces time available for infective agents to spread to healthy herd mates
  • Reduces broad spectrum antibiotic treatment
  • Rapid treatment shortens inflammation time

Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample. Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples, and bulk tank milk samples.

Achieve high sensitivity, even for samples that show no growth in culture. Accurate detection of viable, dead, and growth-inhibited microorganisms reduces no-growth results.

PathoProof Major-3 Kit identifies:

  1. Mycoplasma bovis
  2. Staphylococcus aureus
  3. Streptococcus agalactiae

Automated sample preparation
Significantly reduce the hands-on time needed when processing larger numbers of samples with the semi-automated Thermo Scientific™ KingFisher™ Flex instrument for magnetic bead-based DNA purification and extraction, along with PathoProof Major-3 KingFisher Kits and qPCR reagents.

Not all products are available for sale in all territories. Please inquire.

Permissible Use: This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications).
For testing of Food and Environmental samples only.
Specifications
DescriptionqPCR reagents for use with PF3050L and PF3050LKF
For Use With (Equipment)For use with Applied Biosystems 7500 qPCR systems. Must be ordered together with PF3050L or PF3050LKF.
Kit TypeMajor-3
No. of Tests384 Tests
Product TypeqPCR Reagents for Microbial Detection
Detection MethodqPCR
For Use With (Application)Food & Beverage
GC-Rich PCR PerformanceHigh
PCR MethodMultiplex qPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
qPCR reagents for use with PF3050L and PF3050LKF