Barnstead™ RO Storage Reservoirs and Pumps Accessories
Actual product may vary
Thermo Scientific™
Barnstead™ RO Storage Reservoirs and Pumps Accessories
Choose from various Thermo Scientific™ Barnstead™ RO Storage Reservoirs and Pumps Accessories for your Barnstead RO water purification system, including distribution pumps and sight glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Voltage
PU902X1Optional Distribution PumpPolyethylene Reservoir, 60 L120 V
16975B1Optional Float SwitchBarnstead RO Storage ReservoirN/A
D2623Optional Sight GlassPolyethylene Reservoir, 100 LN/A
PU902X2Optional Distribution PumpPolyethylene Reservoir, 60 L240 V
TU902X1Optional Sight GlassPolyethylene Reservoir, 60 LN/A
Catalog number PU902X1
Price (USD)
-
Type:
Optional Distribution Pump
For Use With (Equipment):
Polyethylene Reservoir, 60 L
Voltage:
120 V
Choose from various Thermo Scientific™ Barnstead™ RO Storage Reservoirs and Pumps Accessories for your Barnstead RO water purification system, including distribution pumps and sight glass.
Specifications
Voltage120 V
For Use With (Equipment)Polyethylene Reservoir, 60 L
TypeOptional Distribution Pump
Unit SizeEach