ProSpecT™ Microplate Assay Wash Buffer
Thermo Scientific™
ProSpecT™ Microplate Assay Wash Buffer
Rapidly detect specific antigens from human samples with Thermo Scientific™ ProSpecT™ Microplate Assays.
Have Questions?
Catalog NumberQuantity
R24500011L/Bottle
Catalog number R2450001
Price (USD)
-
Quantity:
1L/Bottle
Rapidly detect specific antigens in human specimens using Thermo Scientific™ ProSpecT™ Microplate Assays with the Thermo Scientific ProSpecT Microplate Assay Wash Buffer. Its concentration is 10X.
Manage your resources, increase productivity and improve workflow efficiency with direct, qualitative enzyme immmunoassays that combine excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Concentration10X
DescriptionProSpecT Microplate Assay Wash Buffer
FormatBottle
Quantity1L/Bottle
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C