DynaDrive™ BioProcess Container
Thermo Scientific™
DynaDrive™ BioProcess Container
This bioprocess container (BPC) for the DynaDrive Single-Use Bioreactor (S.U.B.) is made with our proprietary Aegis5-14 film. This film isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH31196.013000 L
SH31195.015000 L
Catalog number SH31196.01
Price (USD)
-
Capacity (Metric):
3000 L
This bioprocess container (BPC) for the DynaDrive Single-Use Bioreactor (S.U.B.) is made with our proprietary Aegis5-14 film. This film is widely used by many global biopharmaceutical companies for all bioprocessing steps. Aegis5-14 film was introduced in 2013 and has a clean profile, with completed BPOG leachable and extraction (L&E) testing guidelines for all components used in DynaDrive BPCs.
Key benefits
We offer both standard and custom BPC configurations. Standard configurations are for a variety of common applications, while custom configurations are for customer-specific applications. The key benefits for our BPCs include:
  • Enhanced sparging technology to maximize efficiency while delivering improved mass transfer performance
  • The latest sensor technology in single-use or autoclavable probes, including:
    • pH—electrochemical single-use or reusable probes, and optical single-use sensor
    • pO2—optical single-use or reusable probes
    • Foam—single-use foam sensor built into BPC
    • Capacitance—single-use sensor or reusable probe
    • Raman—optical probe port into BPC
  • Optional ports for alternating tangential flow (ATF) or tangential flow filtration (TFF)
  • Built-in condenser system for 3,000 and 5,000 L BPCs, which helps extend the life and reduce the quantity of exhaust filters needed
  • Utilization of crossflow sparging technology to assure proper head-space gas mixing during low-volume seed or terminal cell cultures
  • Integrated drive-train mixing technology, utilizing four impellers in the 5,000 L DynaDrive S.U.B. and two impellers in the 3,000 L DynaDrive S.U.B. Users no longer need to load a drive shaft into the BPC, thus reducing time and ceiling height constraints
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Quantity3000 L
Capacity (Metric)3000 L
Tubing StyleSterile
Unit Size3,000 L