2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPX insert, Luer Lock, Ladish Tri-Clover
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPX insert, Luer Lock, Ladish Tri-Clover

The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00154I
also known as SS00154-I
200 L
SS00152I
also known as SS00152-I
50 L
SS00153I
also known as SS00153-I
100 L
Catalog number SS00154I
also known as SS00154-I
Price (USD)
-
Capacity (Metric):
200 L
The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 MPX insert + cap, 1 luer lock body connector + valve, 1 Ladish tri-clover).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: MPX insert with 12.7 mm (1/2 in.) barb + cap, pinch clamp. Tubing: C-Flex 374, length: 152.4 cm (60 in.), ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.)
• Line 2: Luer lock body connection with 4.1 mm (5/32 in.) barb + cap, pinch clamp. Tubing: C-Flex 374, length: 101.6 cm (40 in.), ID x OD: 3.2 x 19.1 mm (0.13 x 0.75 in.)
• Line 3: Ladish tri-clover with 12.7 mm (1/2 in.) barb, pinch clamp. Tubing: C-Flex 374, length: 152.4 cm (60 in.), ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.)

Specifications
Dimensions142.2 cm x 95.3 cm (56 in. x 37.5 in.)
For Use With (Equipment)TK-0200-05
Outer Packaging4 mil. poly bag, One 4-mil. poly bag
Product LineLabtainer
Protective Inner PackagingPoly bags on tubing ends, protective material around rough areas
Quantity5 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)200 L
Film DescriptionASI 26/77
MaterialPolyethylene
Port DetailMPX insert + cap, luer lock body connector, and ladish tri-clovers
Tubing StyleC-Flex
Unit Size200 L
Contents & Storage
Store at room temperature.