Thermo Scientific Chemicals

Sulfato de cadmio, anhidro, ACS, > 99 %, Thermo Scientific Chemicals

Número de catálogo: 011861.14
25 g, Each
Thermo Scientific Chemicals

Sulfato de cadmio, anhidro, ACS, > 99 %, Thermo Scientific Chemicals

Número de catálogo: 011861.14
25 g, Each
Cantidad
Informational:Con fines informativos:Este producto químico puede requerir que obtengamos información adicional para nuestros registros de cumplimiento normativo y químico. Si es necesario, nos pondremos en contacto con usted para obtener esta información una vez que haya realizado su pedido.
Número de catálogo: 011861.14
también denominado 011861-14
Precio (USD)
Precio: 31,40
Su precio:
Cantidad
-

Identificadores químicos

CAS
10124-36-4
IUPAC Name
cadmium(2+) sulfate
Molecular Formula
CdO4S
InChI Key
QCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES
[Cd++].[O-]S([O-])(=O)=O
Assay (unspecified)
99.0% CdSO₄ min
Impurity content
Ca 0.01%
Impurity content
Cl 0.001%
Impurity content
Cu 0.002%
Impurity content
Pb 0.003%

Descripción

Cadmium sulfate is used in pigments and electroplating. It acts as an accelerator in cement formation. It is also employed as lubricants. Further, it is useful for the electroplating of cadmium in electronic circuits. It serves as a precursor to prepare cadmium-based pigment like cadmium sulfide. In the Western standard cell, it is used for electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text