Colesteril pelargonato, 99+ %, Thermo Scientific Chemicals
Colesteril pelargonato, 99+ %, Thermo Scientific Chemicals
Colesteril pelargonato, 99+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Colesteril pelargonato, 99+ %, Thermo Scientific Chemicals

Have Questions?
Número de catálogoCantidadEnvase
11027025025 gFrasco de vidrio
Número de catálogo 110270250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1182-66-7
IUPAC Name9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl nonanoate
Molecular FormulaC36H62O2
InChI KeyWCLNGBQPTVENHV-UHFFFAOYNA-N
SMILESCCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Specific optical rotation-28° to -32° (20°C, 589 nm) (C=2, CHCl3)
TLC>=99.0 %
Solubilityalmost colorless
Appearance (Color)White to almost white
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only