Dietil acetilsuccinato, 99 %, Thermo Scientific Chemicals
Dietil acetilsuccinato, 99 %, Thermo Scientific Chemicals
Dietil acetilsuccinato, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dietil acetilsuccinato, 99 %, Thermo Scientific Chemicals

CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
500 g
Envase:
Frasco de vidrio
Número de catálogo 113955000
Precio (USD)
-
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1115-30-6
IUPAC Name1,4-diethyl 2-acetylbutanedioate
Molecular FormulaC10H16O5
InChI KeyDVSDDICSXBCMQJ-UHFFFAOYNA-N
SMILESCCOC(=O)CC(C(C)=O)C(=O)OCC
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only