Etil indol-2-carboxilato, 97 %, Thermo Scientific Chemicals
Etil indol-2-carboxilato, 97 %, Thermo Scientific Chemicals
Etil indol-2-carboxilato, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Etil indol-2-carboxilato, 97 %, Thermo Scientific Chemicals

CAS: 3770-50-1 | C11H11NO2 | 189.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 118600050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS3770-50-1
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder or crystals
GC>=96.0 %
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Melting point119°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only