SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal, 500 rxns
Invitrogen™

SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal, 500 rxns

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoN.º de reaccionesIncluye
11900112500 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control, ezDNase Master Mix and Nuclease-Free Water
1190000150 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control and Nuclease-Free Water
11900002500 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control and Nuclease-Free Water
119000032000 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control and Nuclease-Free Water
1190011150 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control, ezDNase Master Mix and Nuclease-Free Water
119001132000 ReactionsSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control, ezDNase Master Mix and Nuclease-Free Water
119000041250 ReactionsSuperScript V VILO Master Mix, Primer-Free
Número de catálogo 11900112
Precio (USD)
-
N.º de reacciones:
500 Reactions
Incluye:
SuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control, ezDNase Master Mix and Nuclease-Free Water

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy. Powered by SuperScript™ V Reverse Transcriptase, the optimized master mix combines robust performance across a broad range of RNA samples with simplified reaction setup, enhanced reagent stability, and greater workflow flexibility. SuperScript V VILO Master Mix, Primer-Free is also available with ezDNase™ Master Mix for fast and convenient genomic DNA removal prior to reverse transcription.

Features of SuperScript V VILO Master Mix, Primer-Free include:

  • Broad RNA input range—supports sensitive cDNA synthesis from 0.1 pg to 5 μg of total RNA
  • Robust performance with challenging RNA—efficient cDNA synthesis from highly structured, GC-rich, degraded and inhibitor-containing RNA
  • Fast cDNA synthesis—complete reverse transcription in as little as 1-10 minutes
  • Enhanced reagent stability—supports ambient shipping and room-temperature storage for up to 1 month
  • Simple, flexible workflow—ready-to-use format simplifies reaction setup, while room-temperature assembly and extended stability of assembled reactions enable greater workflow flexibility

SuperScript V VILO Master Mix, Primer-Free combines SuperScript V RT, RNase inhibitor, buffer, and dNTPs in a single formulation while allowing users to select and optimize primers for their specific application. This flexibility supports gene-specific primers, oligo(dT), random primers, or customized primer combinations while maintaining the convenience of a ready-to-use master mix.

SuperScript V VILO Master Mix, Primer-Free with gDNA Removal allows streamlined genomic DNA removal step. The dsDNA-specific ezDNase enzyme removes contaminating genomic DNA in as little as 2 minutes without affecting RNA.

SuperScript V VILO Master Mix, Primer-Free also features a DTT-free, animal origin–free formulation for improved sustainability and simplified reagent handling.

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Tipo de producto finalFirst-Strand cDNA
FormatoMaster Mix
IncluyeSuperScript V VILO Master Mix (Primer-Free), supplied with No RT Control, ezDNase Master Mix and Nuclease-Free Water
Inhibitor Tolerance (Simple)High
N.º de reacciones500 Reactions
Temperatura óptima de reacción50°
Tipo de productoReverse Transcription Reagent
Tipo de reactivoReverse Transcription
Transcriptasa inversaSuperScript V
Actividad de la ribonucleasa HReduced
Sensibilidad0.1 pg total RNA
Condiciones de envíoDry Ice, Wet Ice, Ambient
Tamaño (producto final)Up to 20 kb
Material de partidaRNA
TécnicaReverse Transcription
TermoestabilidadUp to 65°
Concentración5X
Método de detecciónqPCR, NGS, PCR
Para utilizar con (aplicación)Gene Expression, Real Time PCR (qPCR), RT-PCR, cDNA Synthesis, Next-Generation Sequencing (NGS)
FormularioLiquid
GC-Rich PCR PerformanceHigh
Etiqueta o tinteNot Applicable
Método de PCR2-step RT-qPCR, RT-PCR
Velocidad de reacción10 min.
Unit SizeEach
Contenido y almacenamiento
• SuperScript V VILO Master Mix, Primer-Free (2 x 1 mL)
• SuperScript V VILO No RT Control, Primer-Free (1 mL)
• ezDNase Master Mix (500 μL)
• Nuclease-free water (1.25 mL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 5 months
• Room temperature for up to 1 month