Thermo Scientific Chemicals

3‘-Cloroacetofenona, 98 %, Thermo Scientific Chemicals

Número de catálogo: 134841000
100 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

3‘-Cloroacetofenona, 98 %, Thermo Scientific Chemicals

Número de catálogo: 134841000
100 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 134841000
Precio (USD)

Identificadores químicos

CAS
99-02-5
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5480 to 1.5500 (20°C, 589 nm)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text