Thermo Scientific Chemicals

Pentafluorobenzaldehído, 98 %, Thermo Scientific Chemicals

Número de catálogo: 147090500
50 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Pentafluorobenzaldehído, 98 %, Thermo Scientific Chemicals

Número de catálogo: 147090500
50 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 147090500
Precio (ARS)

Identificadores químicos

CAS
653-37-2
IUPAC Name
2,3,4,5,6-pentafluorobenzaldehyde
Molecular Formula
C7HF5O
InChI Key
QJXCFMJTJYCLFG-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C=O)C(F)=C1F
Infrared spectrum
Conforms
Refractive index
1.4490 to 1.4510 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid after melting

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text