Ácido gluconico, sal sódica, 98 %, Thermo Scientific Chemicals
Ácido gluconico, sal sódica, 98 %, Thermo Scientific Chemicals
Ácido gluconico, sal sódica, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido gluconico, sal sódica, 98 %, Thermo Scientific Chemicals

CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
1kg
2,5 kg
10 kg
Envase:
Ver todo
Frasco de plástico
Número de catálogo 181390010
Precio (USD)
-
Cantidad:
1kg
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS527-07-1
IUPAC Namesodium 2,3,4,5,6-pentahydroxyhexanoate
Molecular FormulaC6H11NaO7
InChI KeyUPMFZISCCZSDND-UHFFFAOYNA-M
SMILES[Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to light beige
Titration with HClO4>=97.5 %
Chloride (Cl)=<100 ppm
Reducing substances=<0.5 %
Appearance (Form)Crystalline powder
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only