12-Corona-4, 97 %, Thermo Scientific Chemicals
12-Corona-4, 97 %, Thermo Scientific Chemicals
12-Corona-4, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

12-Corona-4, 97 %, Thermo Scientific Chemicals

CAS: 294-93-9 | C8H16O4 | 176.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2.5 g
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 187020050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS294-93-9
IUPAC Name1,4,7,10-tetraoxacyclododecane
Molecular FormulaC8H16O4
InChI KeyXQQZRZQVBFHBHL-UHFFFAOYSA-N
SMILESC1COCCOCCOCCO1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid after melting
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4600 to 1.4630 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactivo de quelación/complexación
RUO – Research Use Only