Ácido D(+)-fenilláctico, 98 %, Thermo Scientific Chemicals
Ácido D(+)-fenilláctico, 98 %, Thermo Scientific Chemicals
Ácido D(+)-fenilláctico, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido D(+)-fenilláctico, 98 %, Thermo Scientific Chemicals

CAS: 7326-19-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 344580010
Precio (USD)
-
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS7326-19-4
IUPAC Name(2R)-2-hydroxy-3-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyVOXXWSYKYCBWHO-MRVPVSSYSA-N
SMILESO[C@H](CC1=CC=CC=C1)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point121°C to 125°C
Titration with NaOH>=97.5 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only