Product Image
Applied Biosystems™
Variant Reporter™ Software v1.0
The Variant Reporter™ Software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing and resequencing with DNAMás información
Have Questions?
Número de catálogoCantidad
44408461 programa
Número de catálogo 4440846
Precio (USD)
-
Cantidad:
1 programa
The Variant Reporter™ Software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing and resequencing with DNA sequencing files. The software is designed for reference based and non-reference based analysis such as mutation detection and analysis, SNP discovery and validation and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletion and heterozygous insertions⁄deletions for data generated using the Applied Biosystems Genetic Analyzers.
• Handle greater that 10 times more sample files than competing software products
• Reduce labor and project cost with accelerated data review
• Improve your results with fast, accurate heterozygote calling
• Simple to learn – requires less than one hour
• Robust data filtering with new more effective quality metrics
Save Time
Manually reviewing your resequencing results requires valuable time and reduces productivity. Variant Reporter™ Software v1.0 does it all for you, quickly, efficiently, and accurately. Designed for basic and clinical researchers engaged in genotyping projects, as well as core resequencing labs whose focus is secondary analysis, this new variant detection software solves the workflow bottleneck caused by the 3–8 hours often required by other software products for data analysis and review. And with your resequencing workflow fully automated, you are free to concentrate on other aspects of your project.
Simple Workflow
To set up a project, simply import and group traces into amplicons and specimens. The analysis parameters can be adjusted and a reference defined, although the data can be analyzed with or without a reference. The software automatically groups traces into specimens and amplicons based on sample file name parsing. Next, analyze your project. After analyzing the data, the software calculates the project-quality statistics and generates an overview of the project data quality. If samples pass QC thresholds, the software will skip this step. Users can also drill down to review the quality of specimens, amplicons, or individual traces. The software contains all the viewing and reporting functionalities of Sequence Scanner Software.
A Choice of Reports
Users can select from the following reports and exports:
• Project Summary Report: Contains high-level project statistics, lists of variants and genotypes, and snippets of each variant
• Quality Report: Provides a comprehensive view of trace, specimen, amplicons, and project quality
• Specimen Report: For clinical researchers interested in reporting data one subject at a time, this report provides specimen statistics, specimen genotypes, and snippets of each variant for one or all specimens in a project.
Use in Resequencing Applications
Resequencing – also known as comparative, direct, medical, or PCR sequencing – is useful for discovering variants, including HIMs, insertions, deletions, SNPs, and genotypes. The process begins when a researcher chooses a gene of interest, isolates DNA from the gene, amplifies one or more amplicons for each exon, and then performs cycle sequencing using the BigDye™ Terminator Cycle Sequencing Kit, followed by cleanup with the BigDye™ XTerminator™ Purification Kit. The sample is then run on an Applied Biosystems capillary electrophoresis instrument. During the final step, Variant Reporter Software discovers the variants, determines the genotypes, and creates a report. That’s all there is to it.
For Research Use Only. Not for use in diagnostics procedures.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Introducción de datosSequencing traces
Salidas de datosVariant Reports with sequence comparison metrics
Para utilizar con (equipo)310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3730 DNA Analyzer
LicenciaStatic
Sistema operativoWindows Vista, Windows XP, Windows Vista Business, Windows 2000 (Service Pack 4), Windows XP (Service Pack 2), Windows 2000
Tipo de productoVariant Reporter Software V1.0
Cantidad1 programa
Categoría de softwareSoftware
Línea de productosVariant Reporter
Software TypeVariant Reporter™ Software, Comparative Sequencing
Unit Size1 software
Preguntas frecuentes
We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?
The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):
-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?
Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up the Data Store in the Variant Reporter Software?
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.