Qualification Services Plan for QuantStudio™ 12K Flex Real-Time PCR System
El producto real puede variar
Applied Biosystems™
Qualification Services Plan for QuantStudio™ 12K Flex Real-Time PCR System
Have Questions?
Número de catálogo 4471980
Precio (USD)
-
Qualification services for the Applied Biosystems QuantStudio 12K Flex Real-Time PCR systems are designed to help balance business and laboratory requirements and help you comply with global standards, country-specific regulations, and other laboratory site requirements.
Verify instrument performance and minimize downtime with IQ/OQ/IPV services
Complete with audit-ready documentation, our factory-trained and experienced engineers will conduct installation qualification (IQ), operational qualification (OQ), and instrument performance verification (IPV) on the qPCR systems in your lab. These qualification services help ensure your instruments are installed, operating, and performing according to manufacturer’s specifications and help move you into production faster.
When are IQ/OQ/IPV recommended?
• At installation
• After a move
When are OQ/IPV recommended?
• After planned maintenance
• After critical repairs
• After additions, changes, or upgrades to the system
• For a scheduled OQ site requirement
For more detailed information, view the figure, "Maintaining a qualified Applied Biosystems real-time PCR (qPCR) instrument" below and visit instrument IQ/OQ/IPV.
Certain exclusions apply. Contact your Services and Support Representative for details at compliancesales@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
For Use With (Equipment)QuantStudio™ 12k Flex
Product LineQuantStudio™
Product SizeIQ/OQ/IPV, 384-well
Unit SizeIQ/OQ/IPV, 384-well