Vanquish™ Active Pre-Heaters TQ
Thermo Scientific™
Vanquish™ Active Pre-Heaters TQ
Contribute to improved resolution and retention time stability in the Thermo Scientific™ Vanquish™ UHPLC system with active or passive eluent pre-heaters using Thermo Scientific™ Viper™ TQ UHPLC Fitting technology.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoLongitud (métrico)
6732.0710380 mm
6732.0700380 mm
6732.0720610 mm
Número de catálogo 6732.0710
Precio (USD)
-
Longitud (métrico):
380 mm
Ensure precise chromatographic performance in the Thermo Scientific™ Vanquish™ UHPLC system with the Thermo Scientific™ Active Pre-heater TQ or Passive Pre-heater TQ. The pre-heaters come with torque-controlled Viper TQ UHPLC Fittings for confident, leak-free connections with audible and tactile feedback to be installed in Thermo Scientific™ Vanquish™ Column Compartment H or C.
The Active Pre-heater matches the temperature of solvent entering the column with the temperature of the column thereby avoiding performance loss from temperature effects. The Active Pre-heater TQ has separate power, heating coil, and temperature controls and comes with replaceable outlet capillaries to extend the lifespan of the part.
Optimizing chromatographic performance with eluent pre-heating
  • Easy to install with the Thermo Scientific™ Viper™ TQ UHPLC fitting technology
  • Reduced dispersion and optimal heat transfer with dedicated formats tailored for the Vanquish HPLC and UHPLC flow paths
  • Wide application flexibility from HPLC to UHPLC using different materials and geometric dimensions
Active pre-heater TQ
The active pre-heater has separate power and a heating coil. The independent temperature control allows the fine-tuning of the eluent temperature for reliable chromatographic methods, method development or method transfer. The replaceable outlet capillaries reduce cost-of-ownership and enhance sustainability with the Active pre-heater TQ beside offering versatile column compatibility.
Passive pre-heater
The passive pre-heater allows for heating and cooling of the eluent to match the column temperature.
Especificaciones
Longitud (métrico)380 mm
MaterialCoated Stainless Steel
TipoPre-heater
Unit SizeEach