Especificaciones
Certificaciones/conformidadEN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15
Dimensiones (Al x An x F)36.1 x 62.5 x 66 cm (14.2 x 24.6 x 26 in.)
Tipo de pantallaFull Color Touch Screen Interface
Frecuencia50/60 Hz
MaterialFront, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
ModeloIVD-MD
Peso neto94 kg / 207 lbs
Nivel de ruido<52 dB(A)
Tipo de productoThermo Scientific X2TR General Purpose Centrifuge
Perfil (aceleración/frenado)9 accel/10 decel rates
Memoria para programasUp to 100 Programs, including multi-step programming via Touch Screen; Including customizable Pulse options
Rotor17 rotor options available, select rotor separately
Rotor incluidoNo
Tiempo de actividadMax 99h, 59min, 59sec; also with continuous and customizable pulse
EstándaresISO 14971, ISO 13485, ISO 9001, FDA listed, Device Class 1
Intervalo de temperatura (métrico)-20°C to +40°C
Ventilado/aNo
Tensión220/230 V
Garantía5 Year Limited (2 years standard warranty plus 3 additional years on motor and refrigeration)
Capacidad4 x 400 mL BioBottles, 76 x 5/7 mL Blood Tubes, 56 x 9/10 mL Blood Tubes, 36 x 15 mL Conical Tubes, 16 x 50 mL Conical Tubes with TX-400 Rotor
RCF máx.25,830 x g
Vel. máx15,200 rpm
RefrigeradoYes
Unit SizeEach