Search
Search

| Número de catálogo | Dimensiones (Al x An x F) | Modelo | Tensión |
|---|---|---|---|
| 75009380 | 36,2 x 62,5 x 66 cm (14,2 x 24,6 x 26 pulg) | IVD | 208/230 V |
| 75009381 | 14,2 x 24,6 x 26 in | IVD | 120 V |
| 75009383 | 14,2 x 24,6 x 26 in | IVD | 110 V |
| 75009880 | 36,2 x 62,5 x 66 cm (14,2 x 24,6 x 26 pulg) | Solo para uso general | 208/230 V |
| 75009883 | 14,2 x 24,6 x 26 in | Certificado IVD | 110 V |
The ST1/ST1R Plus series centrifuges come standard with an easy-to-use keypad and high contrast LCD display. Quickly and securely swap between 14 unique rotor options with the tool-less Auto-Lock rotor exchange. This compact centrifuge has a capacity up to 1.6L and can spin up to 76 x 5/7 mL blood tubes and 16 x 50mL conical tubes with biocontainment and is ideal for routine applications.
Easy to Use:
Thermo Scientific™ Auto-Lock™ III Rotor System:
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
Thermo Scientific™ SMARTSpin™ Technology:
Space-Saving Design:
Energy Efficient:
Explore our clinical centrifuge portfolio for diverse clinical applications.
Learn more