Product Image
Thermo Scientific™

FMDV-Asia Monoclonal Antibody

Monoclonal antibody FMDV-Asia is directed against serotype FMDV Asia 1. The antibody is suited for ELISA and Western blot.
Have Questions?
Número de catálogoCantidad
75009931 mg
Número de catálogo 7500993
Precio (USD)
-
Cantidad:
1 mg

Monoclonal antibody FMDV-Asia is directed against serotype FMDV Asia 1. The antibody is suited for ELISA and Western blot.

Especificaciones
Host abrvMs
Línea de productosPrionics
ReactividadAll species
Reactividad abrvAll
ClasificaciónMonoclonal
Especie del huéspedMouse
Cantidad1 mg
Unit SizeEach