Search
| Número de catálogo | LengthMetric |
|---|---|
| 850-009 | |
| 850-016 |
True average velocity measurement with MASP Technology. MACE velocity sensors use continuous wave Doppler ultrasound to measure the speed of dirt, bubbles and other particles in the stream flow.
The Doppler Ultrasonic Insert Velocity Sensor combines Doppler technology with a unique insertion design that enables operation in pipes from 100 mm to 2,500 mm with the same sensor and installation costs.
This can easily save thousands on installations in large pipes while delivering the accuracy and reliability you need. Install and maintain without shutting down pipe flow, and keep your total cost of ownership low with trouble-free operation, No calibration and no breakable moving parts mean your system is virtually maintenance free.
Install and maintain without shutting down pipe flow, and keep your total cost of ownership low with trouble-free operation, No calibration and no breakable moving parts mean your system is virtually maintenance free. The polycarbonate ZX Snapstrap is tough, secure, does not foul and can be easily removed to take the sensor to another site
For use in full pipes or partially full pipes when used in conjunction with an EchoFlo depth sensor. Insertion sensors measure velocity only and require access to the outside wall of the pipe in which the sensor is to be mounted. These sensors can be installed into existing pipework through a 2 in. ball valve (recommended) or just through a 2 inch female thread fitting.
Technical Specifications