Ácido de hidrazida isonicotínico, + 98 %, Thermo Scientific Chemicals
Ácido de hidrazida isonicotínico, + 98 %, Thermo Scientific Chemicals
Ácido de hidrazida isonicotínico, + 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido de hidrazida isonicotínico, + 98 %, Thermo Scientific Chemicals

CAS: 54-85-3 | C6H7N3O | 137.142 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
50 g
100 g
500 g
1000 g
Número de catálogo A10583.22
también denominado A10583-22
Precio (USD)
-
Cantidad:
100 g
Pedido a granel o personalizado
Identificadores químicos
CAS54-85-3
IUPAC Namepyridine-4-carbohydrazide
Molecular FormulaC6H7N3O
InChI KeyQRXWMOHMRWLFEY-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=NC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to pale cream
FormPowder or crystals
Assay from Suppliers CofA≥98.0% (GC, U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Elemental AnalysisC: 52.55 ± 0.6% (non-U.S. specification)
Ver más
Used for fluorescent HPLC detection of delta 4-3-ketosteroids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only