Thermo Scientific Chemicals

Hexanofenona, 98 %, Thermo Scientific Chemicals

Número de catálogo: A14712.14
25 g, Each
Thermo Scientific Chemicals

Hexanofenona, 98 %, Thermo Scientific Chemicals

Número de catálogo: A14712.14
25 g, Each
Cantidad
Número de catálogo: A14712.14
también denominado A14712-14
Precio (USD)

Identificadores químicos

CAS
942-92-7
IUPAC Name
1-phenylhexan-1-one
Molecular Formula
C12H16O
InChI Key
MAHPVQDVMLWUAG-UHFFFAOYSA-N
SMILES
CCCCCC(=O)C1=CC=CC=C1
Appearance (Color)
Colorless to white to pale yellow
Form
Fused solid or clear liquid as melt
Assay (GC)
≥97.5%
Refractive Index
1.5090-1.5130 @ 20?C

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text